Learn More
ABclonal Technology MCL1 Rabbit pAb
SDP
Supplier:  ABclonal Technology A0434500UL
Encompass_Preferred
This gene encodes an anti-apoptotic protein, which is a member of the Bcl-2 family. Alternative splicing results in multiple transcript variants. The longest gene product (isoform 1) enhances cell survival by inhibiting apoptosis while the alternatively spliced shorter gene products (isoform 2 and isoform 3) promote apoptosis and are death-inducing.
Catalog No. 50-001-37989