Learn More
ABclonal Technology NUMA1 Rabbit pAb
SDP
Supplier:  ABclonal Technology A05271000UL
Encompass_Preferred
This gene encodes a large protein that forms a structural component of the nuclear matrix. The encoded protein interacts with microtubules and plays a role in the formation and organization of the mitotic spindle during cell division. Chromosomal translocation of this gene with the RARA (retinoic acid receptor, alpha) gene on chromosome 17 have been detected in patients with acute promyelocytic leukemia. Alternative splicing results in multiple transcript variants.
Catalog No. 50-001-34687