Learn More
ABclonal Technology EIF3A Rabbit pAb
SDP
Supplier:  ABclonal Technology A05731000UL
Encompass_Preferred
Enables RNA binding activity. Contributes to translation initiation factor activity. Involved in IRES-dependent viral translational initiation, formation of cytoplasmic translation initiation complex, and viral translational termination-reinitiation. Located in cytosol, nucleolus, and nucleoplasm. Part of eukaryotic translation initiation factor 3 complex.
Catalog No. 50-001-35392