Learn More
ABclonal Technology EIF3A Rabbit pAb
SDP
Catalog No. 5000138823
Encompass_Preferred
Enables RNA binding activity. Contributes to translation initiation factor activity. Involved in IRES-dependent viral translational initiation, formation of cytoplasmic translation initiation complex, and viral translational termination-reinitiation. Located in cytosol, nucleolus, and nucleoplasm. Part of eukaryotic translation initiation factor 3 complex.
Catalog No. 50-001-38823 Supplier ABclonal Technology Supplier No. A0573500UL
Enables RNA binding activity. Contributes to translation initiation factor activity. Involved in IRES-dependent viral translational initiation, formation of cytoplasmic translation initiation complex, and viral translational termination-reinitiation. Located in cytosol, nucleolus, and nucleoplasm. Part of eukaryotic translation initiation factor 3 complex.