Learn More
ABclonal Technology EEA1 Rabbit pAb
SDP
Catalog No. 5000136589
Encompass_Preferred
Enables 1-phosphatidylinositol binding activity, GTP-dependent protein binding activity, and protein homodimerization activity. Involved in endocytosis, vesicle fusion, and viral RNA genome replication. Located in cytosol and early endosome. Is extrinsic component of plasma membrane.
Catalog No. 50-001-36589 Supplier ABclonal Technology Supplier No. A0592500UL
Enables 1-phosphatidylinositol binding activity, GTP-dependent protein binding activity, and protein homodimerization activity. Involved in endocytosis, vesicle fusion, and viral RNA genome replication. Located in cytosol and early endosome. Is extrinsic component of plasma membrane.