Learn More
ABclonal Technology Endo G Rabbit pAb
SDP
Supplier:  ABclonal Technology A0604500UL
Encompass_Preferred
The protein encoded by this gene is a nuclear encoded endonuclease that is localized in the mitochondrion. The encoded protein is widely distributed among animals and cleaves DNA at GC tracts. This protein is capable of generating the RNA primers required by DNA polymerase gamma to initiate replication of mitochondrial DNA.
Catalog No. 50-001-35380