Learn More
ABclonal Technology GSK3α Rabbit pAb
SDP
Supplier:  ABclonal Technology A0645500UL
Encompass_Preferred
This gene encodes a multifunctional Ser/Thr protein kinase that is implicated in the control of several regulatory proteins including glycogen synthase, and transcription factors, such as JUN. It also plays a role in the WNT and PI3K signaling pathways, as well as regulates the production of beta-amyloid peptides associated with Alzheimers disease.
Catalog No. 50-001-36603