Learn More
ABclonal Technology ANGPTL3 Rabbit pAb
SDP
Supplier:  ABclonal Technology A06891000UL
Encompass_Preferred
This gene encodes a member of a family of secreted proteins that function in angiogenesis. The encoded protein, which is expressed predominantly in the liver, is further processed into an N-terminal coiled-coil domain-containing chain and a C-terminal fibrinogen chain. The N-terminal chain is important for lipid metabolism, while the C-terminal chain may be involved in angiogenesis. Mutations in this gene cause familial hypobetalipoproteinemia type 2.
Catalog No. 50-001-38360