Learn More
ABclonal Technology TGF beta Receptor I (TGFBR1) Rabbit pAb
SDP
Supplier:  ABclonal Technology A0708500UL
Encompass_Preferred
The protein encoded by this gene forms a heteromeric complex with type II TGF-beta receptors when bound to TGF-beta, transducing the TGF-beta signal from the cell surface to the cytoplasm. The encoded protein is a serine/threonine protein kinase. Mutations in this gene have been associated with Loeys-Dietz aortic aneurysm syndrome (LDAS). Multiple transcript variants encoding different isoforms have been found for this gene.
Catalog No. 50-001-37931