Learn More
ABclonal Technology CD34 Rabbit pAb
SDP
Supplier:  ABclonal Technology A0761500UL
Encompass_Preferred
The protein encoded by this gene may play a role in the attachment of stem cells to the bone marrow extracellular matrix or to stromal cells. This single-pass membrane protein is highly glycosylated and phosphorylated by protein kinase C. Two transcript variants encoding different isoforms have been found for this gene.
Catalog No. 50-001-38022