Learn More
ABclonal Technology cIAP1 Rabbit pAb
SDP
Supplier:  ABclonal Technology A0866500UL
Encompass_Preferred
The protein encoded by this gene is a member of a family of proteins that inhibits apoptosis by binding to tumor necrosis factor receptor-associated factors TRAF1 and TRAF2, probably by interfering with activation of ICE-like proteases. This encoded protein inhibits apoptosis induced by serum deprivation and menadione, a potent inducer of free radicals. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Catalog No. 50-001-38671