Learn More
ABclonal Technology NSF Rabbit pAb
SDP
Catalog No. 5000136705
Encompass_Preferred
Enables PDZ domain binding activity and ionotropic glutamate receptor binding activity. Involved in intracellular protein transport, positive regulation of protein catabolic process, and positive regulation of receptor recycling. Located in Golgi apparatus, cytosol, and plasma membrane. Implicated in developmental and epileptic encephalopathy.
Catalog No. 50-001-36705 Supplier ABclonal Technology Supplier No. A0926500UL
Enables PDZ domain binding activity and ionotropic glutamate receptor binding activity. Involved in intracellular protein transport, positive regulation of protein catabolic process, and positive regulation of receptor recycling. Located in Golgi apparatus, cytosol, and plasma membrane. Implicated in developmental and epileptic encephalopathy.