Learn More
ABclonal Technology SFPQ Rabbit pAb
SDP
Supplier:  ABclonal Technology A09581000UL
Encompass_Preferred
Enables DNA binding activity, histone deacetylase binding activity, and protein homodimerization activity. Involved in several processes, including alternative mRNA splicing, via spliceosome, positive regulation of oxidative stress-induced intrinsic apoptotic signaling pathway, and regulation of transcription by RNA polymerase II. Acts upstream of or within double-strand break repair via homologous recombination. Located in chromatin, nuclear matrix, and paraspeckles.
Catalog No. 50-001-35003