Learn More
ABclonal Technology SFPQ Rabbit pAb
SDP
Catalog No. 5000135702
Encompass_Preferred
Enables DNA binding activity, histone deacetylase binding activity, and protein homodimerization activity. Involved in several processes, including alternative mRNA splicing, via spliceosome, positive regulation of oxidative stress-induced intrinsic apoptotic signaling pathway, and regulation of transcription by RNA polymerase II. Acts upstream of or within double-strand break repair via homologous recombination. Located in chromatin, nuclear matrix, and paraspeckles.
Catalog No. 50-001-35702 Supplier ABclonal Technology Supplier No. A0958500UL
Enables DNA binding activity, histone deacetylase binding activity, and protein homodimerization activity. Involved in several processes, including alternative mRNA splicing, via spliceosome, positive regulation of oxidative stress-induced intrinsic apoptotic signaling pathway, and regulation of transcription by RNA polymerase II. Acts upstream of or within double-strand break repair via homologous recombination. Located in chromatin, nuclear matrix, and paraspeckles.