Learn More
ABclonal Technology γ-Catenin Rabbit pAb
SDP
Supplier:  ABclonal Technology A09631000UL
Encompass_Preferred
This gene encodes a major cytoplasmic protein which is the only known constituent common to submembranous plaques of both desmosomes and intermediate junctions. This protein forms distinct complexes with cadherins and desmosomal cadherins and is a member of the catenin family since it contains a distinct repeating amino acid motif called the armadillo repeat. Mutation in this gene has been associated with Naxos disease. Alternative splicing occurs in this gene, however, not all transcripts have been fully described.
Catalog No. 50-001-38078