Learn More
ABclonal Technology NEK8 Rabbit pAb
SDP
Supplier:  ABclonal Technology A09841000UL
Encompass_Preferred
This gene encodes a member of the serine/threionine protein kinase family related to NIMA (never in mitosis, gene A) of Aspergillus nidulans. The encoded protein may play a role in cell cycle progression from G2 to M phase. Mutations in the related mouse gene are associated with a disease phenotype that closely parallels the juvenile autosomal recessive form of polycystic kidney disease in humans.
Catalog No. 50-001-35261