Learn More
New England Biolabs, Inc. α1-2,3,6 Mannosidase - 80 units
SDP
Catalog No. 501526427
Encompass
1-2,3,6 Mannosidase, cloned from Jack Bean, and also known as JBM, is a broad specificity exoglycosidase that catalyzes the hydrolysis of terminal 1-2, 1-3 and 1-6 linked mannose residues from oligosaccharides. 1-2,3,6 Mannosidase has a slight preference for 1-2 mannose residues over 1-3 and 1-6 mannose residues.
Catalog No. 50-152-6427 Supplier New England Biolabs, Inc. Supplier No. P0768S
1-2,3,6 Mannosidase, cloned from Jack Bean, and also known as JBM, is a broad specificity exoglycosidase that catalyzes the hydrolysis of terminal 1-2, 1-3 and 1-6 linked mannose residues from oligosaccharides. 1-2,3,6 Mannosidase has a slight preference for 1-2 mannose residues over 1-3 and 1-6 mannose residues.