Learn More
New England Biolabs, Inc. α1-2,4,6 Fucosidase O - 80 units
SDP
Catalog No. 501526425
Encompass
a1-2,4,6 Fucosidase O is a broad specificity exoglycosidase that catalyzes the hydrolysis of terminal a1-2, a1-4 and a1-6 linked fucose residues from oligosaccharides. a1-2,4,6 Fucosidase O cleaves a1-6 fucose residues more efficiently than other linkages.
Catalog No. 50-152-6425 Supplier New England Biolabs, Inc. Supplier No. P0749S
a1-2,4,6 Fucosidase O is a broad specificity exoglycosidase that catalyzes the hydrolysis of terminal a1-2, a1-4 and a1-6 linked fucose residues from oligosaccharides. a1-2,4,6 Fucosidase O cleaves a1-6 fucose residues more efficiently than other linkages.