Learn More
New England Biolabs, Inc. α1-3,4,6 Galactosidase - 200 units
SDP
Catalog No. 501526423
Encompass
a1-3,4,6 Galactosidase is a broad specificity exoglycosidase that catalyzes the hydrolysis of terminal, non-reducing a1-3, a1-4 and a1-6-linked galactose residues from oligosaccharides.
Catalog No. 50-152-6423 Supplier New England Biolabs, Inc. Supplier No. P0747S
a1-3,4,6 Galactosidase is a broad specificity exoglycosidase that catalyzes the hydrolysis of terminal, non-reducing a1-3, a1-4 and a1-6-linked galactose residues from oligosaccharides.