Learn More
Athens Research And Technology Inc A1 ACID GLYCOPROTEIN HU 10MG
Supplier:  Athens Research And Technology Inc 161601070010MG
Encompass_Preferred
Alpha 1 Acid Glycoprotein, Human Plasma: 10 MG; Salt-free lyophilized solid; MW 40,000; Extinction Coefficient = 0.89; Purity: > =95% by SDS-PAGE; This native glycoprotein purified from human plasma in the Athens Research & Tech lab.
Catalog No. 50-489-600