Learn More
Athens Research And Technology Inc Alpha 1 Antitrypsin, Human Plasma | 5mg
Catalog No. NC1329697
Encompass_Preferred
An acute-phase plasma protein found at 95-350 mg per 100 ml that functions as a protease inhibitor. Clinically, its deficiency is associated with two major diseases: pulmonary emphysema and early onset/juvenile hepatic cirrhosis. It is elevated in inflammatory conditions, malignancies, liver disease and pregnancy and also after surgical trauma and use of oral contraceptives. The simultaneous quantitative determination of alpha-1-PI and ceruloplasmin permits differential diagnosis of liver afflictions.
Catalog No. NC1329697 Supplier Athens Research And Technology Inc Supplier No. 16160116095MG
Add to Cart
Add to Cart
An acute-phase plasma protein found at 95-350 mg per 100 ml that functions as a protease inhibitor. Clinically, its deficiency is associated with two major diseases: pulmonary emphysema and early onset/juvenile hepatic cirrhosis. It is elevated in inflammatory conditions, malignancies, liver disease and pregnancy and also after surgical trauma and use of oral contraceptives. The simultaneous quantitative determination of alpha-1-PI and ceruloplasmin permits differential diagnosis of liver afflictions.