Learn More
BD FACSymphony™ A1 Cell Analyzer
Catalog No. BDB664893 Shop All Waters Corporation Products
Change view
Click to view available options
Includes:
In-Person Training
Catalog No. Includes
BDB664893 In-Person Training
1 options
Catalog No. BDB664893 Supplier BD FACSymphony™ Supplier No. 664893
Please contact your sales representative for a quote or further ordering instructions.
The FACSymphony™ A1 Cell Analyzer is a premium performance flow cytometry cell analyzer in a benchtop footprint.
The BD FACSymphony™ A1 Cell Analyzer features high-end technology from BD FACSymphony™ platforms in a compact size. The BD FACSymphony™ A1 Cell Analyzer supports high-resolution analysis of 16 fluorescent markers simultaneously, by utilizing low-noise electronics, proprietary optical design and high-powered lasers.
  • Offers premium high-end BD FACSymphony™ instrument technology scaled to fit on your benchtop
  • Utilizes industry standard BD FACSDiva™ Software for streamlined workflow from system setup to data acquisition and analysis
  • Is compatible with the broad portfolio of catalog and BD RealBlue™ and RealYellow™ Reagents and BD FACSDiva™ Software to streamline your flow cytometry workflow
  • Permits easy placement on a standard lab bench with its compact format
TRUSTED_SUSTAINABILITY
Specifications
Laser Features Four 100 mW Lasers (Violet 405 nm, Blue 488 nm, Yellow-Green 561 nm, Red 637 nm)
For Use With (Equipment) BD FACSDiva™ Software for streamlined workflow from system setup to data acquisition and analysis
Voltage 100/240 VAC
Includes In-Person Training