Learn More
Cayman Chemical a1-MIcroglobulInhumnreco 100ug
SDP
Catalog No. 502916361
Encompass_Preferred
Source: Recombinant human C-terminal His-tagged α1-microglobulin expressed in HEK293 cells • Amino acids: 20-203 • MW: 22.3 kDa
Catalog No. 50-291-6361 Supplier Cayman Chemical Supplier No. 38053100 UG
Source: Recombinant human C-terminal His-tagged α1-microglobulin expressed in HEK293 cells • Amino acids: 20-203 • MW: 22.3 kDa