Learn More
ATCC A-10; Embryonic Smooth Muscle; Rat (Rattus norvegicus)
Catalog No. 502383492
Encompass
A-10 is a myoblast-like cell isolated from the thoracic aorta of a rat. This cell line was deposited by W Carlisle and can be used for cardiovascular disease research.
Catalog No. 50-238-3492 Supplier ATCC Supplier No. CRL1476
Add to Cart
Add to Cart
A-10 is a myoblast-like cell isolated from the thoracic aorta of a rat. This cell line was deposited by W Carlisle and can be used for cardiovascular disease research.