Learn More
Miele, Inc. A 100 Upper Basket for Modules
Catalog No. NC1892280
Encompass
Upper basket with two docking pipes. For the connection of up to two injector modules or inserts. Automatically self-sealing docking valves.
Catalog No. NC1892280 Supplier Miele, Inc. Supplier No. 09862320
Upper basket with two docking pipes. For the connection of up to two injector modules or inserts. Automatically self-sealing docking valves.