Learn More
ABclonal Technology KLF6 Rabbit pAb
SDP
Catalog No. 5000138454
Encompass_Preferred
This gene encodes a member of the Kruppel-like family of transcription factors. The zinc finger protein is a transcriptional activator, and functions as a tumor suppressor. Multiple transcript variants encoding different isoforms have been found for this gene, some of which are implicated in carcinogenesis.
Catalog No. 50-001-38454 Supplier ABclonal Technology Supplier No. A10011500UL
This gene encodes a member of the Kruppel-like family of transcription factors. The zinc finger protein is a transcriptional activator, and functions as a tumor suppressor. Multiple transcript variants encoding different isoforms have been found for this gene, some of which are implicated in carcinogenesis.