Learn More
ABclonal Technology PAPD7 Rabbit pAb
SDP
Catalog No. 5000138114
Encompass_Preferred
The protein encoded by this gene is a DNA polymerase that is likely involved in DNA repair. In addition, the encoded protein may be required for sister chromatid adhesion. Alternatively spliced transcript variants that encode different isoforms have been described.
Catalog No. 50-001-38114 Supplier ABclonal Technology Supplier No. A100171000UL
The protein encoded by this gene is a DNA polymerase that is likely involved in DNA repair. In addition, the encoded protein may be required for sister chromatid adhesion. Alternatively spliced transcript variants that encode different isoforms have been described.