Learn More
ABclonal Technology [KO Validated] LRRC59 Rabbit pAb
SDP
Catalog No. 5000134622
Encompass_Preferred
Enables RNA binding activity and cadherin binding activity. Predicted to be involved in positive regulation of Ras protein signal transduction and signal transduction. Located in endoplasmic reticulum and mitochondrial nucleoid.
Catalog No. 50-001-34622 Supplier ABclonal Technology Supplier No. A10022500UL
Enables RNA binding activity and cadherin binding activity. Predicted to be involved in positive regulation of Ras protein signal transduction and signal transduction. Located in endoplasmic reticulum and mitochondrial nucleoid.