Learn More
ABclonal Technology NOSIP Rabbit pAb
SDP
Catalog No. 5000138215
Encompass_Preferred
The protein encoded by this gene may modulate the activity and localization of nitric oxide synthase (endothelial and neuronal) and thus nitric oxide production. Alternative splicing results in multiple transcript variants that encode the same protein.
Catalog No. 50-001-38215 Supplier ABclonal Technology Supplier No. A10024500UL
The protein encoded by this gene may modulate the activity and localization of nitric oxide synthase (endothelial and neuronal) and thus nitric oxide production. Alternative splicing results in multiple transcript variants that encode the same protein.