Learn More
ABclonal Technology RBP1 Rabbit pAb
SDP
Supplier:  ABclonal Technology A100291000UL
Encompass_Preferred
This gene encodes the carrier protein involved in the transport of retinol (vitamin A alcohol) from the liver storage site to peripheral tissue. Vitamin A is a fat-soluble vitamin necessary for growth, reproduction, differentiation of epithelial tissues, and vision. Multiple transcript variants encoding different isoforms have been found for this gene.
Catalog No. 50-001-38214