Learn More
ABclonal Technology [KO Validated] UBE2O Rabbit pAb
SDP
Catalog No. 5000138472
Encompass_Preferred
Enables ubiquitin conjugating enzyme activity and ubiquitin protein ligase activity. Involved in positive regulation of BMP signaling pathway, protein ubiquitination, and retrograde transport, endosome to Golgi. Located in cytoplasm and nuclear body.
Catalog No. 50-001-38472 Supplier ABclonal Technology Supplier No. A100361000UL
Add to Cart
Add to Cart
Enables ubiquitin conjugating enzyme activity and ubiquitin protein ligase activity. Involved in positive regulation of BMP signaling pathway, protein ubiquitination, and retrograde transport, endosome to Golgi. Located in cytoplasm and nuclear body.