Learn More
ABclonal Technology [KO Validated] UBE2O Rabbit pAb
SDP
Supplier:  ABclonal Technology A10036500UL
Encompass_Preferred
Enables ubiquitin conjugating enzyme activity and ubiquitin protein ligase activity. Involved in positive regulation of BMP signaling pathway, protein ubiquitination, and retrograde transport, endosome to Golgi. Located in cytoplasm and nuclear body.
Catalog No. 50-001-38136