Learn More
ABclonal Technology RPRD1B Rabbit pAb
SDP
Catalog No. 5000136672
Encompass_Preferred
Enables RNA polymerase II complex binding activity and identical protein binding activity. Involved in positive regulation of cell population proliferation, regulation of cell cycle process, and regulation of transcription by RNA polymerase II. Located in nucleoplasm. Part of RNA polymerase II, holoenzyme.
Catalog No. 50-001-36672 Supplier ABclonal Technology Supplier No. A10102500UL
Enables RNA polymerase II complex binding activity and identical protein binding activity. Involved in positive regulation of cell population proliferation, regulation of cell cycle process, and regulation of transcription by RNA polymerase II. Located in nucleoplasm. Part of RNA polymerase II, holoenzyme.