Learn More
ABclonal Technology SLC31A1 Rabbit pAb
SDP
Supplier:  ABclonal Technology A101091000UL
Encompass_Preferred
The protein encoded by this gene is a high-affinity copper transporter found in the cell membrane. The encoded protein functions as a homotrimer to effect the uptake of dietary copper.
Catalog No. 50-001-36671