Learn More
ABclonal Technology SLC31A1 Rabbit pAb
SDP
Catalog No. 5000135316
Encompass_Preferred
The protein encoded by this gene is a high-affinity copper transporter found in the cell membrane. The encoded protein functions as a homotrimer to effect the uptake of dietary copper.
Catalog No. 50-001-35316 Supplier ABclonal Technology Supplier No. A10109500UL
The protein encoded by this gene is a high-affinity copper transporter found in the cell membrane. The encoded protein functions as a homotrimer to effect the uptake of dietary copper.