Learn More
ABclonal Technology INPPL1 Rabbit pAb
SDP
Supplier:  ABclonal Technology A101151000UL
Encompass_Preferred
The protein encoded by this gene is an SH2-containing 5-inositol phosphatase that is involved in the regulation of insulin function. The encoded protein also plays a role in the regulation of epidermal growth factor receptor turnover and actin remodelling. Additionally, this gene supports metastatic growth in breast cancer and is a valuable biomarker for breast cancer.
Catalog No. 50-001-37588