Learn More
ABclonal Technology NDUFA10 Rabbit pAb
SDP
Supplier:  ABclonal Technology A101231000UL
Encompass_Preferred
The protein encoded by this gene is a component of 42 kDa complex I, the first enzyme complex in the electron transport chain of mitochondria. This protein has NADH dehydrogenase activity and oxidoreductase activity. It transfers electrons from NADH to the respiratory chain. A mutation in this gene was found in an individual with Leigh syndrome.
Catalog No. 50-001-36686