Learn More
ABclonal Technology TRIB1 Rabbit pAb
SDP
Catalog No. 5000135320
Encompass_Preferred
Enables mitogen-activated protein kinase kinase binding activity and protein kinase inhibitor activity. Involved in several processes, including JNK cascade, negative regulation of lipopolysaccharide-mediated signaling pathway, and regulation of protein kinase activity. Located in cytoplasm and nucleus.
Catalog No. 50-001-35320 Supplier ABclonal Technology Supplier No. A101341000UL
Enables mitogen-activated protein kinase kinase binding activity and protein kinase inhibitor activity. Involved in several processes, including JNK cascade, negative regulation of lipopolysaccharide-mediated signaling pathway, and regulation of protein kinase activity. Located in cytoplasm and nucleus.