Learn More
ABclonal Technology Casein Kinase 2 alpha (CSNK2A1) Rabbit pAb
SDP
Supplier:  ABclonal Technology A10141000UL
Encompass_Preferred
Casein kinase II is a serine/threonine protein kinase that phosphorylates acidic proteins such as casein. It is involved in various cellular processes, including cell cycle control, apoptosis, and circadian rhythm. The kinase exists as a tetramer and is composed of an alpha, an alpha-prime, and two beta subunits. The alpha subunits contain the catalytic activity while the beta subunits undergo autophosphorylation. The protein encoded by this gene represents the alpha subunit. Multiple transcript variants encoding different protein isoforms have been found for this gene.
Catalog No. 50-001-35910