Learn More
ABclonal Technology TRPM4 Rabbit pAb
SDP
Catalog No. 5000135120
Encompass_Preferred
The protein encoded by this gene is a calcium-activated nonselective ion channel that mediates transport of monovalent cations across membranes, thereby depolarizing the membrane. The activity of the encoded protein increases with increasing intracellular calcium concentration, but this channel does not transport calcium.
Catalog No. 50-001-35120 Supplier ABclonal Technology Supplier No. A101461000UL
The protein encoded by this gene is a calcium-activated nonselective ion channel that mediates transport of monovalent cations across membranes, thereby depolarizing the membrane. The activity of the encoded protein increases with increasing intracellular calcium concentration, but this channel does not transport calcium.