Learn More
ABclonal Technology TRPM4 Rabbit pAb
SDP
Supplier:  ABclonal Technology A10146500UL
Encompass_Preferred
The protein encoded by this gene is a calcium-activated nonselective ion channel that mediates transport of monovalent cations across membranes, thereby depolarizing the membrane. The activity of the encoded protein increases with increasing intracellular calcium concentration, but this channel does not transport calcium.
Catalog No. 50-001-35912