Learn More
ABclonal Technology GRHL2 Rabbit pAb
SDP
Catalog No. 5000138674
Encompass_Preferred
The protein encoded by this gene is a transcription factor that can act as a homodimer or as a heterodimer with either GRHL1 or GRHL3. Defects in this gene are a cause of non-syndromic sensorineural deafness autosomal dominant type 28 (DFNA28).
Catalog No. 50-001-38674 Supplier ABclonal Technology Supplier No. A10153500UL
The protein encoded by this gene is a transcription factor that can act as a homodimer or as a heterodimer with either GRHL1 or GRHL3. Defects in this gene are a cause of non-syndromic sensorineural deafness autosomal dominant type 28 (DFNA28).