Learn More
ABclonal Technology TAS1R3 Rabbit pAb
SDP
Catalog No. 5000136691
Encompass_Preferred
The protein encoded by this gene is a G-protein coupled receptor involved in taste responses. The encoded protein can form a heterodimeric receptor with TAS1R1 to elicit the umami taste response, or it can bind with TAS1R2 to form a receptor for the sweet taste response.
Catalog No. 50-001-36691 Supplier ABclonal Technology Supplier No. A10157500UL
The protein encoded by this gene is a G-protein coupled receptor involved in taste responses. The encoded protein can form a heterodimeric receptor with TAS1R1 to elicit the umami taste response, or it can bind with TAS1R2 to form a receptor for the sweet taste response.