Learn More
ABclonal Technology PPP1R14A Rabbit pAb
SDP
Supplier:  ABclonal Technology A101601000UL
Encompass_Preferred
The protein encoded by this gene belongs to the protein phosphatase 1 (PP1) inhibitor family. This protein is an inhibitor of smooth muscle myosin phosphatase, and has higher inhibitory activity when phosphorylated. Inhibition of myosin phosphatase leads to increased myosin phosphorylation and enhanced smooth muscle contraction. Alternatively spliced transcript variants encoding different isoforms have been noted for this gene.
Catalog No. 50-001-36510