Learn More
ABclonal Technology C6 Rabbit pAb
SDP
Catalog No. 5000135851
Encompass_Preferred
This gene encodes a component of the complement cascade. The encoded protein is part of the membrane attack complex that can be incorporated into the cell membrane and cause cell lysis. Mutations in this gene are associated with complement component-6 deficiency. Transcript variants encoding the same protein have been described.
Catalog No. 50-001-35851 Supplier ABclonal Technology Supplier No. A101701000UL
This gene encodes a component of the complement cascade. The encoded protein is part of the membrane attack complex that can be incorporated into the cell membrane and cause cell lysis. Mutations in this gene are associated with complement component-6 deficiency. Transcript variants encoding the same protein have been described.