Learn More
ABclonal Technology CNPase Rabbit pAb
SDP
Catalog No. 5000135130
Encompass_Preferred
Predicted to enable 2,3-cyclic-nucleotide 3-phosphodiesterase activity. Involved in substantia nigra development. Located in several cellular components, including extracellular space, microtubule, and plasma membrane. Implicated in hypomyelinating leukodystrophy 20, multiple sclerosis, and schizophrenia. Biomarker of alcoholic liver cirrhosis, multiple sclerosis, and restless legs syndrome.
Catalog No. 50-001-35130 Supplier ABclonal Technology Supplier No. A1018500UL
Predicted to enable 2,3-cyclic-nucleotide 3-phosphodiesterase activity. Involved in substantia nigra development. Located in several cellular components, including extracellular space, microtubule, and plasma membrane. Implicated in hypomyelinating leukodystrophy 20, multiple sclerosis, and schizophrenia. Biomarker of alcoholic liver cirrhosis, multiple sclerosis, and restless legs syndrome.