Learn More
ABclonal Technology SMPD3 Rabbit pAb
SDP
Catalog No. 5000135131
Encompass_Preferred
Predicted to enable phosphatidic acid binding activity, phosphatidylserine binding activity, and sphingomyelin phosphodiesterase activity. Predicted to be involved in positive regulation of exosomal secretion and sphingomyelin metabolic process. Predicted to act upstream of or within several processes, including animal organ development, enzyme linked receptor protein signaling pathway, and sphingolipid metabolic process. Predicted to be located in Golgi apparatus and plasma membrane. Predicted to be active in cytoplasm. Biomarker of pulmonary emphysema.
Catalog No. 50-001-35131 Supplier ABclonal Technology Supplier No. A101971000UL
Predicted to enable phosphatidic acid binding activity, phosphatidylserine binding activity, and sphingomyelin phosphodiesterase activity. Predicted to be involved in positive regulation of exosomal secretion and sphingomyelin metabolic process. Predicted to act upstream of or within several processes, including animal organ development, enzyme linked receptor protein signaling pathway, and sphingolipid metabolic process. Predicted to be located in Golgi apparatus and plasma membrane. Predicted to be active in cytoplasm. Biomarker of pulmonary emphysema.