Learn More
ABclonal Technology SMPD3 Rabbit pAb
SDP
Supplier:  ABclonal Technology A10197500UL
Encompass_Preferred
Predicted to enable phosphatidic acid binding activity, phosphatidylserine binding activity, and sphingomyelin phosphodiesterase activity. Predicted to be involved in positive regulation of exosomal secretion and sphingomyelin metabolic process. Predicted to act upstream of or within several processes, including animal organ development, enzyme linked receptor protein signaling pathway, and sphingolipid metabolic process. Predicted to be located in Golgi apparatus and plasma membrane. Predicted to be active in cytoplasm. Biomarker of pulmonary emphysema.
Catalog No. 50-001-36541