Learn More
ABclonal Technology DIAPH2 Rabbit pAb
SDP
Catalog No. 5000138570
Encompass_Preferred
The product of this gene belongs to the diaphanous subfamily of the formin homology family of proteins. This gene may play a role in the development and normal function of the ovaries. Defects in this gene have been linked to premature ovarian failure 2. Alternatively spliced transcript variants encoding different isoforms have been identified.
Catalog No. 50-001-38570 Supplier ABclonal Technology Supplier No. A102091000UL
The product of this gene belongs to the diaphanous subfamily of the formin homology family of proteins. This gene may play a role in the development and normal function of the ovaries. Defects in this gene have been linked to premature ovarian failure 2. Alternatively spliced transcript variants encoding different isoforms have been identified.