Learn More
ABclonal Technology DIAPH2 Rabbit pAb
SDP
Supplier:  ABclonal Technology A10209500UL
Encompass_Preferred
The product of this gene belongs to the diaphanous subfamily of the formin homology family of proteins. This gene may play a role in the development and normal function of the ovaries. Defects in this gene have been linked to premature ovarian failure 2. Alternatively spliced transcript variants encoding different isoforms have been identified.
Catalog No. 50-001-36712